Learn More
Analytical Sales & Services HTE Kit 1, for 24-Well Screening
Catalog No. 5000805171
Encompass
Contains:\nA) Four (4) 24-well Block Assemblies\na1) 2 packs of Top PFA Film, 24-well (25/pack)\na2) 1 pack of Bottom PFA Film, 24-well (25/pack)\na3) 1 pack of Top Red Mats, 24-well (25/pack)\na4) 1 pack of Bottom Red Mats, 24-well (25/pack)\nD) Spare Screws\nE) Electric Screwdriver\nF) Case of 1000 1mL 8x30 Shell Vials\nI) Pack of 1000 Tumble Stir Elements
Catalog No. 50-008-05171 Supplier Analytical Sales & Services Supplier No. HTEKIT1
Contains:\nA) Four (4) 24-well Block Assemblies\na1) 2 packs of Top PFA Film, 24-well (25/pack)\na2) 1 pack of Bottom PFA Film, 24-well (25/pack)\na3) 1 pack of Top Red Mats, 24-well (25/pack)\na4) 1 pack of Bottom Red Mats, 24-well (25/pack)\nD) Spare Screws\nE) Electric Screwdriver\nF) Case of 1000 1mL 8x30 Shell Vials\nI) Pack of 1000 Tumble Stir Elements