Learn More
Promocell GmbH PromoCell HTEpC-c 500,000 Human Tracheal Epithelial Cells, cryopreserved
Supplier:  Promocell GmbH C12644
Encompass
Primary Human Tracheal Epithelial Cells (HTEpC) are isolated from the surface epithelium of human trachea and stain positive for cytokeratin.The respiratory epithelia are responsible for the lubrication of the lungs, the maintenance of humidity, and the cleaning of the respiratory tract. They are an important target for drugs, toxins, and carcinogens. In addition, they are involved in many diseases such as cystic fibrosis. Consequently, HTEpC are useful for investigating the function and pathology of the respiratory system.Our HTEpC are now also available from HLA-typed donors.Available formats:* Cryopreserved: Cryogenic vial containing 500.000 viable cells.* Proliferating: >500.000 viable cells shipped in growth medium (T25 flask).* Cell pellet: 1 million cells dissolved in 200µl RNAlater© for subsequent RNA, DNA or protein analysis. Cell pellets cannot be revived.
Catalog No. 50-306-050