Learn More
Alstem hTERT Cell Immortalization Kit 5 x 20ul
SDP
Catalog No. NC0999989
Encompass
hTERT Cell Immortalization Kit - Puromycin Simply infect your cells with our recombinant lentivirus containing hTERT and screen with puromycin to generate your own immortalized cells. 5 x 20ul
Catalog No. NC0999989 Supplier Alstem Supplier No. CILV02
hTERT Cell Immortalization Kit - Puromycin Simply infect your cells with our recombinant lentivirus containing hTERT and screen with puromycin to generate your own immortalized cells. 5 x 20ul