Learn More
Alstem HTERT-Neo 5 x 20ul
SDP
Catalog No. NC1851239
Encompass
hTERT Cell Immortalization Kit - Neomycin Simply infect your cells with our recombinant lentivirus containing hTERT and screen with neomycin to generate your own immortalized cells. 5 x 20ul
Catalog No. NC1851239 Supplier Alstem Supplier No. CILV04
hTERT Cell Immortalization Kit - Neomycin Simply infect your cells with our recombinant lentivirus containing hTERT and screen with neomycin to generate your own immortalized cells. 5 x 20ul