Learn More
Gentarget HTERT EF1A GFP-PURO LENTIVIRUS
Catalog No. NC3312350
Encompass
Pre-made Lentivirus expressing Human Telomerase Reverse Transcriptase /hTERT NM_198253.2/ under enhanced EF1a promoter. A GFP-Puromycin fusion dual marker was expressed under RSV promoter, which allows to select the transduced cell via Puromycin antibiotic or via cell sorting by GFP fluorescent signal. hTERT is widely used for cell immortalization used alone or together with other cell immortalization factors like SV40 large T-antigen.
Catalog No. NC3312350 Supplier Gentarget Supplier No. LVP1131GP
Pre-made Lentivirus expressing Human Telomerase Reverse Transcriptase /hTERT NM_198253.2/ under enhanced EF1a promoter. A GFP-Puromycin fusion dual marker was expressed under RSV promoter, which allows to select the transduced cell via Puromycin antibiotic or via cell sorting by GFP fluorescent signal. hTERT is widely used for cell immortalization used alone or together with other cell immortalization factors like SV40 large T-antigen.