Learn More
Gentarget LVP1131-GP-PBS, pre-made lentivirus for hTERT expression, 1 x 10^8 IFU/ml, 200 μl/vial, PBS solution, GFP-Puromycin selection, Enhanced EF1a promoter
Catalog No. NC3467135
Encompass
LVP1131-GP-PBS, pre-made lentivirus for hTERT expression, 1 x 10^8 IFU/ml, 200 μl/vial, PBS solution, GFP-Puromycin selection, Enhanced EF1a promoter
Catalog No. NC3467135 Supplier Gentarget Supplier No. LVP1131GPPBS
LVP1131-GP-PBS, pre-made lentivirus for hTERT expression, 1 x 10^8 IFU/ml, 200 μl/vial, PBS solution, GFP-Puromycin selection, Enhanced EF1a promoter