Learn More
ATCC hTERT Immortalized RPTEC Growth Kit
Catalog No. 502383754
Encompass
hTERT Immortalized RPTEC Growth Kit can be added to DMEM: F12 (ATCC 30-2006) to produce a complete medium for hTERT Immortalized RPTEC cells. The kit consists of individually packaged supplements; each supplement must be added in appropriate volumes as per Product Sheet directions to 500 ml basal medium, DMEM: F12.
Catalog No. 50-238-3754 Supplier ATCC Supplier No. ACS4007
hTERT Immortalized RPTEC Growth Kit can be added to DMEM: F12 (ATCC 30-2006) to produce a complete medium for hTERT Immortalized RPTEC cells. The kit consists of individually packaged supplements; each supplement must be added in appropriate volumes as per Product Sheet directions to 500 ml basal medium, DMEM: F12.