Learn More
ATCC hTERT Mesenchymal; Stem Cell Exosomes; Human (Homo sapiens)
Catalog No. 502383513
Encompass
Exosomes are subcellular nanoparticles of size 50-200 nm released from cells through a fusion of multicellular bodies with plasma membrane. The exosomes are purified from cell culture medium of hTERT-immortalized Mesenchymal Stem Cells (ATCC SCRC-4000). The exosomes are purified and concentrated using tangential flow filtration system in a sterile environment. The purified exosomes were further filtered through 0.2 micron sterile filters.
Catalog No. 50-238-3513 Supplier ATCC Supplier No. SCRC4000EXM
Exosomes are subcellular nanoparticles of size 50-200 nm released from cells through a fusion of multicellular bodies with plasma membrane. The exosomes are purified from cell culture medium of hTERT-immortalized Mesenchymal Stem Cells (ATCC SCRC-4000). The exosomes are purified and concentrated using tangential flow filtration system in a sterile environment. The purified exosomes were further filtered through 0.2 micron sterile filters.