Learn More
TRIBIOSCIENCE INC Human Tubal Fluid (HTF) Mouse Embryo Medium, 50 ML
SDP
Catalog No. NC3454605
Encompass_Preferred
Tribioscience’s Human Tubal Fluid (HTF) medium is intended for use mouse embryo cultures, which include gamete and embryo manipulation and embryo culture. This medium is a synthetic, defined solution for use as an embryo culture media. The Size of medium is 50 mL/ bottle.
Catalog No. NC3454605 Supplier TRIBIOSCIENCE INC Supplier No. TBS8072
Tribioscience’s Human Tubal Fluid (HTF) medium is intended for use mouse embryo cultures, which include gamete and embryo manipulation and embryo culture. This medium is a synthetic, defined solution for use as an embryo culture media. The Size of medium is 50 mL/ bottle.