Learn More
Julabo USA Inc DD-BC4 heating circulator, 1kW heating power, 4L bath, to 200C, 115V/60Hz
SDP
Catalog No. 501539900
Encompass_Preferred
JULABO DYNEO™ DD-BC4 Heating circulator +20¿+200 ¿C ¿0.01 ¿C; 1 kW heater; 4 L bath tank; Bath opening (WxL/D): 5.1x5.0/5.9 inches; adjustable flow pump 8-23L/min; 1.5-8.7 psi with 8 and 12 mm hose barbs; for internal/external applications; Color TFT display with central rotary knob for input. Integrated programmer. Multi-lingual. USB port for PC communication; Pt100 port for external temperature control; Dimensions (WxLxH): 9.1x16.1x15.5 inches; 19 pounds. 115V/60Hz.
Catalog No. 50-153-9900 Supplier Julabo USA Inc Supplier No. 9021504.22
JULABO DYNEO™ DD-BC4 Heating circulator +20¿+200 ¿C ¿0.01 ¿C; 1 kW heater; 4 L bath tank; Bath opening (WxL/D): 5.1x5.0/5.9 inches; adjustable flow pump 8-23L/min; 1.5-8.7 psi with 8 and 12 mm hose barbs; for internal/external applications; Color TFT display with central rotary knob for input. Integrated programmer. Multi-lingual. USB port for PC communication; Pt100 port for external temperature control; Dimensions (WxLxH): 9.1x16.1x15.5 inches; 19 pounds. 115V/60Hz.