Learn More
Apexbio Technology LLC HTH-01-015(Synonyms: HTH01015, WZ4003, NUAK1 inhibitor WZ4003, NUAK1-IN-1, NUAK1 inhibitor, WZ-4003), 10mM (in 1mL DMSO), CAS: 1613724-42-7.
SDP
Catalog No. 5000060057
Encompass_Preferred
HTH-01-015 (CAS 1613724-42-7) is a highly selective inhibitor targeting the NUAK1 kinase exhibiting potent inhibitory activity with an IC50 of approximately 100 nM It specifically blocks NUAK1-mediated phosphorylation of MYPT1 at Ser445 with negligible off-target effects among a comprehensive panel of 139 other kinases Cellular studies demonstrate that treatment with HTH-01-015 reduces migration of mouse embryonic fibroblasts (MEFs) suppresses proliferation in U2OS and MEF cell lines at concentrations around 10 M and significantly inhibits invasion of U2OS cells within 3D Matrigel transwell assays This compound serves as a useful research tool for investigating NUAK1-related signaling pathways in cancer biology and cellular dynamics
Catalog No. 50-000-60057 Supplier Apexbio Technology LLC Supplier No. A88905.1
HTH-01-015 (CAS 1613724-42-7) is a highly selective inhibitor targeting the NUAK1 kinase exhibiting potent inhibitory activity with an IC50 of approximately 100 nM It specifically blocks NUAK1-mediated phosphorylation of MYPT1 at Ser445 with negligible off-target effects among a comprehensive panel of 139 other kinases Cellular studies demonstrate that treatment with HTH-01-015 reduces migration of mouse embryonic fibroblasts (MEFs) suppresses proliferation in U2OS and MEF cell lines at concentrations around 10 M and significantly inhibits invasion of U2OS cells within 3D Matrigel transwell assays This compound serves as a useful research tool for investigating NUAK1-related signaling pathways in cancer biology and cellular dynamics