Learn More
Medchemexpress LLC Hth-01-015 | 1613724-42-7 | 98.8% | 468.55 | C26H28N8O | 10MG
SDP
Catalog No. 5000238083
Encompass_Preferred
HTH-01-015 is a selective small-molecule inhibitor of NUAK1 (ARK5) with an in vitro IC50 of 100 nM and >100-fold selectivity over NUAK2. Supplied as a solid for research use, it is intended for biochemical and cellular studies probing NUAK1 signaling pathways.
  • Selective NUAK1 inhibition with an IC50 of 100 nM.
  • Greater than 100-fold selectivity versus NUAK2.
  • High reported purity (~98.8%) suitable for research use.
  • Available in multiple pack sizes, including milligram quantities.
  • Supplied as a solid with recommended cold storage for stability.
  • Includes analytical documents (COA, LC-MS, 1H NMR) for quality verification.
Catalog No. 50-002-38083 Supplier Medchemexpress LLC Supplier No. HY1233410MG
HTH-01-015 is a selective small-molecule inhibitor of NUAK1 (ARK5) with an in vitro IC50 of 100 nM and >100-fold selectivity over NUAK2. Supplied as a solid for research use, it is intended for biochemical and cellular studies probing NUAK1 signaling pathways.
  • Selective NUAK1 inhibition with an IC50 of 100 nM.
  • Greater than 100-fold selectivity versus NUAK2.
  • High reported purity (~98.8%) suitable for research use.
  • Available in multiple pack sizes, including milligram quantities.
  • Supplied as a solid with recommended cold storage for stability.
  • Includes analytical documents (COA, LC-MS, 1H NMR) for quality verification.