Learn More
Selleck Chemical LLC HTH-01-015 5mg 1613724-42-7
SDP
Supplier:  Selleck Chemical LLC S73185MG
Encompass_Preferred
HTH-01-015 is a potent and selective NUAK1 inhibitor with IC50 of 100 nM, >100-fold selectivity over NUAK2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4639