Learn More
Medchemexpress LLC HY-12334 5mg Medchemexpress, HTH-01-015 CAS:1613724-42-7 Purity:>98%
SDP
Catalog No. 502029956
Encompass_Preferred
Medchemexpress, HY-12334 5mg HTH-01-015 CAS:1613724-42-7 HTH-01-015 is a selective NUAK1/ARK5 inhibitor (IC50 is 100 nM). HTH-01-015 inhibits NUAK1 with >100-fold higher potency than NUAK2 (IC50 of >10 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9956 Supplier Medchemexpress LLC Supplier No. HY123345MG
Add to Cart
Add to Cart
Medchemexpress, HY-12334 5mg HTH-01-015 CAS:1613724-42-7 HTH-01-015 is a selective NUAK1/ARK5 inhibitor (IC50 is 100 nM). HTH-01-015 inhibits NUAK1 with >100-fold higher potency than NUAK2 (IC50 of >10 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.