Learn More
TARGETMOL CHEMICALS INC HTH-01-091 5MG
SDP
Catalog No. 502580827
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HTH-01-091 is a potent and selective maternal embryonic leucine zipper kinase (MELK) inhibitor (IC50 10.5 nM).HTH-01-091 inhibits PIM1/2/3 RIPK2 DYRK3 smMLCK and CLK2. HTH-01-091 can be used to study breast cancer. purity: 98%
Catalog No. 50-258-0827 Supplier TARGETMOL CHEMICALS INC Supplier No. T241525MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HTH-01-091 is a potent and selective maternal embryonic leucine zipper kinase (MELK) inhibitor (IC50 10.5 nM).HTH-01-091 inhibits PIM1/2/3 RIPK2 DYRK3 smMLCK and CLK2. HTH-01-091 can be used to study breast cancer. purity: 98%