Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hth-02-006 | 99.4% | 588.44 | C25H29IN6O3 | 10 MG
SDP

Catalog No. 5000287823
Encompass_Preferred

HTH-02-006 is a small-molecule NUAK2 inhibitor (IC50 = 126 nM) used in cellular and animal models to study YAP-driven liver and prostate cancer. It reduces NUAK2-mediated signaling and has demonstrated growth inhibition in YAP-high cancer cells and anti-tumor efficacy in mouse models.

  • Target: NUAK2
  • IC50 126 nM (NUAK2)
  • Purity 99.4%
  • Molecular formula C25H29IN6O3
  • Molecular weight 588.44
  • Form off-white to light yellow solid
  • Solubility DMSO 100 mg/mL (in vitro)
  • Storage powder -20°C (long term), 4°C (short term); in solvent -80°C (6 months), -20°C (1 month)
  • Pack size 10 mg
  • Suitable for cellular and in vivo cancer research models

Catalog No. 50-002-87823 Supplier Medchemexpress LLC Supplier No. HY15639710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HTH-02-006 is a small-molecule NUAK2 inhibitor (IC50 = 126 nM) used in cellular and animal models to study YAP-driven liver and prostate cancer. It reduces NUAK2-mediated signaling and has demonstrated growth inhibition in YAP-high cancer cells and anti-tumor efficacy in mouse models.

  • Target: NUAK2
  • IC50 126 nM (NUAK2)
  • Purity 99.4%
  • Molecular formula C25H29IN6O3
  • Molecular weight 588.44
  • Form off-white to light yellow solid
  • Solubility DMSO 100 mg/mL (in vitro)
  • Storage powder -20°C (long term), 4°C (short term); in solvent -80°C (6 months), -20°C (1 month)
  • Pack size 10 mg
  • Suitable for cellular and in vivo cancer research models

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.