Learn More
Selleck Chemical LLC HTHQ (1-O-Hexyl-2.3.5-trimethylhydroquinone) S5314-25mg
SDP
Catalog No. 503110437
Encompass_Preferred
HTHQ (1-O-Hexyl-2 3 5-trimethylhydroquinone) which is a hydroquinone monoalkylether is a potent anti-oxidative and anti-lipid-peroxidative agent HTHQ(1-O-Hexyl-2 3 5-trimethylhydroquinone) has considerable anti-oxidative activity by directly reacting with reactive oxygen species (ROS) including peroxyl radicals and scavenging them to form more stable free radicals
Catalog No. 50-311-0437 Supplier Selleck Chemical LLC Supplier No. S531425MG
HTHQ (1-O-Hexyl-2 3 5-trimethylhydroquinone) which is a hydroquinone monoalkylether is a potent anti-oxidative and anti-lipid-peroxidative agent HTHQ(1-O-Hexyl-2 3 5-trimethylhydroquinone) has considerable anti-oxidative activity by directly reacting with reactive oxygen species (ROS) including peroxyl radicals and scavenging them to form more stable free radicals