Learn More
Bioworld HTLV-1 ENVELOPE (RECOMBINANT)
Supplier:  Bioworld 220602241
Encompass_Preferred
]Recombinant HTLV-I Envelope Source:E.coli Description: The protein contains the C- terminus of gp46 and most of p21E of HTLV-I. This non-fusion contains immunodominant fragment ,amino acids (165-440). Purification method: Purified...
Catalog No. 50-253-325