Learn More
Creative Biomart Recombinant HTLV-1 gp21 and gp46 mosaic Antigen, GST-tagged
Supplier:  Creative Biomart HTL252
Encompass_Preferred
The E. coli derived recombinant mosaic protein contains the gp21 and gp46 immunodominant regions, 374-400 amino acids and 190-207 amino acids, the MW is 42 kDa. The protein is fused with GST at N-term
Catalog No. 50-208-4630