Learn More
Bioworld HTLV-1 p24 Core (Recombinant), 100 μg
Supplier:  Bioworld 220602271
Encompass_Preferred
Recombinant HTLV-I core protein p24. Source:E.coli Description : The protein contains the full-length HTLV-I major core protein p24. Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Protein conc: 1.0 mg/ml, in 50 mM Na-PO4. pH 6.0, containing 1 mM DTT and 1 mM EDTA. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Stability : Five years frozen. One month in solution at room temperature. Sfecificity: Immunoreactive with all sera of HTLV-I infected individuals. Applications: Antigen in ELISA and Western blots, excellent antigen for early detection of HIV seroconvertors with minimal specificity problems. Usage: BioWORLD\\\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-328