Learn More
CPC Scientific H-Leu-Leu-Phe-Gly-Tyr-Pro-Val-Tyr-Val-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HTLV001A
Encompass
Human T lymphotropic virus type 1 HTLV-1 Tax (11-19) is an immunodominant HLA-A2-restricted epitope. It is recognized by a high number of CD8+ cytotoxic T lymphocytes (CTLs) in patients with HTLV-1-associated myelopathy/tropical spastic paraparesis (HAM/TSP), a demyelinating inflammatory neurologic disease associated with HTLV-1 infection. H-6096 therefore represents a useful tool to elucidate the role of antigen-specific CD8+ CTLs in HAM/TSP.ONE-LETTER SEQUENCE: LLFGYPVYVMOLECULAR FORMULA: C56H79N9O12MOLECULAR WEIGHT:1070.3STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [141677-18-1], RESEARCH AREA: AntimicrobialREFERENCES: T.F.Greten et al., Proc. Natl. Acad. Sci. USA, 95, 7568 (1998); U.Tomaru et al., Nat. Med., 9, 469 (2003)
Catalog No. 50-210-9579