Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ HTM Agar for Susceptibility Testing
SDP

Catalog No. 23001797 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-797 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-797 Supplier Hardy Diagnostics™ Supplier No. G33
Add to Cart
Add to Cart
This item is not returnable. View return policy

HTM Agar (Haem.Test Med.) for disk diffusion sensitivity testing of Haemophilus spp.

  • HTM is an uncomplicated growth medium that is transparent to facilitate the reading of susceptibility tests

Specifications

Appearance Clear, Slightly Opalescent
Color Medium to Dark Amber
Content And Storage 2°C to 8°C
Country of Origin United States
Additive Hematin, NAD
Detects Antimicrobial Susceptibility in Haemophilus Species
Form Agar
Format Agar Plate
Manufacturing Origin United States
Media Type Agar
Product Type Agar Plate
Target Organism Class Fastidious Organisms
Test Type Antimicrobial Susceptibility
Fill Volume 18 mL
Formulation Acid Hydrolysate of Casein - 17.5gm, Yeast Extract - 5.0gm, Beef Extract - 2.0gm, Starch - 1.5gm, Hematin - 15.0mg, NAD - 15.0mg, Agar - 17.0gm
For Use With (Application) Antimicrobial Disk Susceptibility Testing of Haemophilus Species
Incubation Conditions 35°C
Incubation Time 16 to 18 hr.
Packaging Type Bag
Quantity 25 mL
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Haemophilus influenzae, Escherichia coli
Protein Source Casein, Yeast, Beef
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.