Learn More
Perkin Elmer US LLC HTM Demountable Cell Kit - ZnSe, Qty. 2
Catalog No. 502111676
Encompass_Preferred
The HTM Demountable Cell (L1390046) is a component of the Heatable Transmission Module (L1390029/L1370130), an accessory for Spectrum spectrometers. This cell should be ordered with at least one set of windows (L1390047/L1390048/L1390049).
Catalog No. 50-211-1676 Supplier Perkin Elmer US LLC Supplier No. L1390047
The HTM Demountable Cell (L1390046) is a component of the Heatable Transmission Module (L1390029/L1370130), an accessory for Spectrum spectrometers. This cell should be ordered with at least one set of windows (L1390047/L1390048/L1390049).