Learn More
Abcam HTMT (Histamine trifluoromethyl toluidide) dimaleate, H1/H2 receptor agonist, 50MG
Catalog No. 502706875
Encompass_Preferred
MW 628.6 Da, Purity >99%. H1/H2 receptor agonist (Ki = 1.2 µM). More potent than histamine (ab120734). Activates IP3/Ca2+ signaling and induces proliferation of small cholangiocytes. Active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6875 Supplier Abcam Supplier No. AB14112750MG
MW 628.6 Da, Purity >99%. H1/H2 receptor agonist (Ki = 1.2 µM). More potent than histamine (ab120734). Activates IP3/Ca2+ signaling and induces proliferation of small cholangiocytes. Active *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement