Learn More
Invivogen Anti-hTNF-alpha-hIgG1, 100 ug
SDP
Supplier:  Invivogen HTNFAMAB1
Encompass
Anti-hTNF-alpha-hIgG1 was generated by recombinant DNA technology. It is produced in CHO cells and purified by affinity chromatography with protein G. The neutralizing activity of anti-hTNF-alpha-hIgG1 antibody was determined using HEK-Blue TNF-alpha cells.
Catalog No. NC0231008