Learn More
Research Products International Corp Heating Block Module for 20mm Tubes
SDP
Catalog No. 502229514
Encompass
These precision machined solid aluminum blocks are meant to accommodate micro-centrifuge tubes, microplates and test tubes. Anodized blocks resist corrosion and feature alpha/numerical labeled rows and columns for easy vessel identification. Each block has a well for a thermometer (sold separately) and a block extractor hole.
Block Modules can be used in either an analog or digital dry bath incubator.
Catalog No. 50-222-9514 Supplier Research Products International Corp Supplier No. 110020
These precision machined solid aluminum blocks are meant to accommodate micro-centrifuge tubes, microplates and test tubes. Anodized blocks resist corrosion and feature alpha/numerical labeled rows and columns for easy vessel identification. Each block has a well for a thermometer (sold separately) and a block extractor hole.
Block Modules can be used in either an analog or digital dry bath incubator.