Learn More
Aviva Systems Biology Corp HTR1A Antibody - N-terminal region (AVARP13041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HTR1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HTR1A. Alternate names: 5HT1a, 5-HT1A, ADRBRL1, ADRB2RL1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53784