Learn More
Aviva Systems Biology Corp HTR1F Antibody - N-terminal region (AVARP13073_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13073_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HTR1F. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HTR1F. Alternate names: 5HT6, MR77, 5-HT1F, HTR1EL, 5-HT-1FApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62632