Learn More
Aviva Systems Biology Corp HTR2A Antibody - N-terminal region (AVARP13044_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13044_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HTR2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HTR2A. Alternate names: 5-HT2A, HTR2ApplicationWB. Species reactivity: Human, Mouse, Pig
Catalog No. 50-014-53787