Learn More
Aviva Systems Biology Corp Htr3a Antibody - middle region (ARP35615_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35615_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Htr3a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: 5-HT3, 5-HT3A, 5HT3, Htr3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-284-468