Learn More
Aviva Systems Biology Corp HTR3B Antibody - N-terminal region (ARP35186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HTR3B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human HTR3B. Alternate names: 5-HT3BApplicationIF, IHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55665