Learn More
Aviva Systems Biology Corp HTR3E Antibody - middle region (ARP35532_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35532_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HTR3E. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human HTR3E. Alternate names: 5-HT3c1, MGC120035, MGC120036, MGC120037, 5-HT3E, 5-HT3-EApplicationIF, WB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-014-55774