Learn More
Cusabio Technology LLC HtrA serine peptidase 1
Catalog No. 5000050814
Encompass_Preferred
This kit detects HTRA1 in mouse serum plasma and tissue homogenates with a sensitivity of 19 5 pg/mL offering a quantitative measurement range of 78 pg/mL to 5000 pg/mL Utilizing a sandwich ELISA technique it requires 50-100 L of sample per test and delivers results within 1-5 hours The assay employs an absorbance readout at 450 nm for precise detection ensuring reliable and efficient analysis of HTRA1 protein levels in biological samples Designed for research applications this kit provides a robust tool for studying HTRA1 expression and its role in physiological or pathological processes
Catalog No. 50-000-50814 Supplier Cusabio Technology LLC Supplier No. CSBEL010901MO
This kit detects HTRA1 in mouse serum plasma and tissue homogenates with a sensitivity of 19 5 pg/mL offering a quantitative measurement range of 78 pg/mL to 5000 pg/mL Utilizing a sandwich ELISA technique it requires 50-100 L of sample per test and delivers results within 1-5 hours The assay employs an absorbance readout at 450 nm for precise detection ensuring reliable and efficient analysis of HTRA1 protein levels in biological samples Designed for research applications this kit provides a robust tool for studying HTRA1 expression and its role in physiological or pathological processes