Learn More
Sino Biological Recombinant Human GUCY2F Protein (GST Tag)
Catalog No. 502574807
Encompass_Preferred
Recombinant human GUCY2F (367-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4807 Supplier Sino Biological Supplier No. G1231G100
Recombinant human GUCY2F (367-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.