Learn More
Revvity Health Sciences Inc Sulfur-35 Radionuclide, 5mCi (185MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sodium Sulfate in 1mL Water
Catalog No. 502812842
Encompass
Sulfur-35 Radionuclide, 5mCi (185MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sodium Sulfate in 1mL Water
Catalog No. 50-281-2842 Supplier Revvity Health Sciences Inc Supplier No. NEX041H005MC
Add to Cart
Add to Cart
Sulfur-35 Radionuclide, 5mCi (185MBq), Specific Activity: 1050-1600Ci (38.8-59.2TBq)/mmol, Sodium Sulfate in 1mL Water