Learn More
Revvity Health Sciences Inc HTRF Human Total FGFR3 Detection Kit, 50,000 Assay Points
Catalog No. 502811772
Encompass
HTRF Human Total FGFR3 Detection Kit, 50,000 Assay Points
Catalog No. 50-281-1772 Supplier Revvity Health Sciences Inc Supplier No. 64FGFR3TPEY
Add to Cart
Add to Cart
Request a Quote
HTRF Human Total FGFR3 Detection Kit, 50,000 Assay Points