Learn More
Revvity Health Sciences Inc HTRF Human Total eNOS Detection Kit, 500 Assay Points
Catalog No. NC3918417
Encompass
The Total eNOS kit is designed to monitor the expression level of cellular eNOS (endothelial nitric oxide synthase), and can be used as a normalization assay for the Phospho-eNOS (Ser1177) kit.
Catalog No. NC3918417 Supplier Revvity Health Sciences Inc Supplier No. 64ENOSTPEG
The Total eNOS kit is designed to monitor the expression level of cellular eNOS (endothelial nitric oxide synthase), and can be used as a normalization assay for the Phospho-eNOS (Ser1177) kit.