Learn More
Revvity Health Sciences Inc HTRF Human Total Tie2 Detection Kit, 500 assay points
Catalog No. NC3918061
Encompass
This HTRF kit allows for the cell-based quantitative detection of Total Tie2.
Catalog No. NC3918061 Supplier Revvity Health Sciences Inc Supplier No. 64TIE2TPEG
This HTRF kit allows for the cell-based quantitative detection of Total Tie2.