Learn More
Eurofins Discoverx PrecisION hTRPA1 Recombinant Stable Cell Line, 2 vials. (CYL3066)
Supplier:  Eurofins Discoverx CYL3066
Encompass
The PrecisION hTRPA1 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying TRPA1 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses TRPA1 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/ion-channels.
Catalog No. 50-200-5097
Add to Cart
Request a Quote