Learn More
Perkin Elmer US LLC HTS Internal Standard Port 7 Tubing, 0.75 mm I.D.
Catalog No. 502112567
Encompass_Preferred
This Internal Standard Port 7 Tubing is included in the Sample Delivery Tubing and Loop Kit for HTS (N8150145). PerkinElmers High Throughput System (HTS) affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. Using a metal-free fluid path, the system is able to quickly deliver the sample to plasma, while having uncompromised precision and detection limits.
Catalog No. 50-211-2567 Supplier Perkin Elmer US LLC Supplier No. N8152924
This Internal Standard Port 7 Tubing is included in the Sample Delivery Tubing and Loop Kit for HTS (N8150145). PerkinElmers High Throughput System (HTS) affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. Using a metal-free fluid path, the system is able to quickly deliver the sample to plasma, while having uncompromised precision and detection limits.