Learn More
Perkin Elmer US LLC HTS Waste Tubing, 1.5 mm I.D.
Catalog No. 502112569
Encompass_Preferred
This Waster Tubing is included in the Sample Delivery Tubing and Loop Kit for HTS (N8150145). PerkinElmers High Throughput System (HTS) affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. Using a metal-free fluid path, the system is able to quickly deliver the sample to plasma, while having uncompromised precision and detection limits.
Catalog No. 50-211-2569 Supplier Perkin Elmer US LLC Supplier No. N8152927
This Waster Tubing is included in the Sample Delivery Tubing and Loop Kit for HTS (N8150145). PerkinElmers High Throughput System (HTS) affords ultra-fast analysis, dramatically reducing laboratory overheads and the cost of analysis. Using a metal-free fluid path, the system is able to quickly deliver the sample to plasma, while having uncompromised precision and detection limits.