Learn More
Medchemexpress LLC HY-101503 5mg Medchemexpress, HTS01037 CAS:682741-29-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015035MG
Encompass_Preferred
Medchemexpress, HY-101503 5mg HTS01037 CAS:682741-29-3 HTS01037 is an inhibitor of fatty acid binding; and a competitive antagonist of protein-protein interactions mediated by AFABP/aP2 with a K i of 0.67 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2153