Learn More
Promocell GmbH PromoCell HTSMC-c 500,000 Human Tracheal Smooth Muscle Cells, cryopreserved
Supplier:  Promocell GmbH C12565
Encompass
Primary Human Tracheal Smooth Muscle Cells (HTSMC) are isolated from the trachea from single donors and stain positive for smooth muscle α-actin. They connect the endings of the tracheal cartilage by a contractile bridge called the paries membranaceus and consequently regulate the airway lumen. Under pathological conditions like asthma, the amount of smooth muscle cells in the trachea walls is increased leading to smooth muscle hypertrophy or hyperplasia. Hence, HTSMC are suitable for in vitro studies of asthma and other pulmonary diseases.Our HTSMC are now also available from HLA-typed donors.Available formats:* Cryopreserved: Cryogenic vial containing 500.000 viable cells.* Proliferating: >500.000 viable cells shipped in growth medium (T25 flask).* Cell pellet: 1 million cells dissolved in 200µl RNAlater© for subsequent RNA, DNA or protein analysis. Cell pellets cannot be revived.
Catalog No. 50-306-035