Learn More
Bon Opus Biosciences Recombinant Human ACE-2 (C-Fc) 1mg
SDP
Supplier:  Bon Opus Biosciences C05Y1MG
Encompass_Preferred
Recombinant Human Angiotensin-Converting Enzyme 2 is produced by our Mammalian expression system and the target gene encoding Gln18-Ser740 is expressed with a Fc tag at the C-terminus. Accession# Q9BYF1. Formulation: Supplied as a 0.2 μm filtered solution of 20mM Tris-HCl, 300mM NaCl, 1mM ZnCl2, 10% Glycerol, pH 7.4. Bon Opus Cat# C05Y
Catalog No. 50-217-2719