Learn More
Bon Opus Biosciences Recombinant Human Acrosomal Protein SP-10/ACRV1 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C4211MG
Encompass_Preferred
Recombinant Human Acrosomal Protein SP-10 is produced by our Mammalian expression system and the target gene encoding Gln22-Ile265 is expressed with a 6His tag at the C-terminus. Accession# P26436. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C421
Catalog No. 50-217-2741