Learn More
Bon Opus Biosciences Recombinant Human/Mouse Activin A/INHBA -500ug
SDP
Catalog No. 502080361
Encompass_Preferred
Recombinant Human Activin A is produced by our Mammalian expression system and the target gene encoding Gly311-Ser426 is expressed. Expression system: Human Cells. Accession# P08476. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# C687
Catalog No. 50-208-0361 Supplier Bon Opus Biosciences Supplier No. C687500UG
Recombinant Human Activin A is produced by our Mammalian expression system and the target gene encoding Gly311-Ser426 is expressed. Expression system: Human Cells. Accession# P08476. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# C687