Learn More
Bon Opus Biosciences Recombinant Human Alcohol Dehydrogenase Class 4 Mu/ADH7 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C8821MG
Encompass_Preferred
Recombinant Human ADH7 is produced by our Mammalian expression system and the target gene encoding Met1-Phe386 is expressed with a 6His tag at the C-terminus. Accession# P40394. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS,PH7.4. Bon Opus Cat# C882
Catalog No. 50-217-2760