Learn More
Bon Opus Biosciences Recombinant Human Aldehyde Dehydrogenase 1-A1/ALDH1A1 (N-6His) 10ug
SDP
Supplier:  Bon Opus Biosciences CF4410UG
Encompass_Preferred
Recombinant Human Aldehyde Dehydrogenase 1-A1 is produced by our E.coli expression system and the target gene encoding Met1-Ser501 is expressed with a 6His tag at the N-terminus. Accession# P00352. Formulation: Supplied as a 0.2 μm filtered solution of 20mM TrisHCl, pH8.5. Bon Opus Cat# CF44
Catalog No. 50-217-2784