Learn More
Bon Opus Biosciences Recombinant Human ANGPTL4 (N-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW5350UG
Encompass_Preferred
Recombinant Human Angiopoietin Like Protein 4 is produced by our Mammalian expression system and the target gene encoding Pro166-Ser406 is expressed with a 6His tag at the N-terminus. Expression system: Human Cells. Accession# Q9BY76. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW53
Catalog No. 50-208-0435