Learn More
Bon Opus Biosciences Recombinant Human Annexin A6/ANXA6 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C2071MG
Encompass_Preferred
Recombinant Human Annexin A6 is produced by our E.coli expression system and the target gene encoding Ala2-Asp673 is expressed with a 6His tag at the C-terminus. Accession# P08133. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# C207
Catalog No. 50-217-2727